The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001024468.3 | NP_001019639.1 | 5 - 6 | 749 | 66 |
NM_007532.5 | NP_031558.3 | 5 - 6 | 768 | 66 | |
GenBank mRNA | AK013888.1 | - | 5 - 6 | 705 | 66 |
AK036309.1 | - | 5 - 6 | 750 | 66 | |
AK138468.1 | - | 5 - 6 | 662 | 66 | |
AK166971.1 | - | 5 - 6 | 704 | 66 | |
AK167232.1 | - | 5 - 6 | 695 | 66 | |
AK167368.1 | - | 5 - 6 | 614 | 66 | |
AK168249.1 | - | 5 - 6 | 798 | 66 | |
BC053706.1 | - | 5 - 6 | 700 | 66 | |
U42443.1 | - | 4 - 5 | 505 | 66 | |
X17502.1 | - | 5 - 6 | 692 | 66 |