Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Prpf4
Entrez Gene ID
70052
Gene Name
PRP4 pre-mRNA processing factor 4 homolog (yeast)
Gene Aliases
1600015H11Rik, AI874830, AW047464, bN189G18.1
Chromosome Location
Chr.4: 62408767 - 62426990 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_027297.3 NP_081573.1 13 - 14 1463 70
GenBank mRNA AK005464.1 - 13 - 14 1442 70
AK085174.1 - 13 - 14 1427 70
AK089838.1 - 13 - 14 1415 70
AK090114.1 - 13 - 14 1440 70
AK166912.1 - 13 - 14 1449 70
AK167216.1 - 13 - 14 1441 70
AK169532.1 - 13 - 14 1426 70
BC051639.1 - 9 - 10 877 70
BC106089.1 - 13 - 14 1425 70
BC125529.1 - 13 - 14 1414 70
BC125531.1 - 13 - 14 1414 70

View Full Product Details