Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Wtip
Entrez Gene ID
101543
Gene Name
WT1-interacting protein
Gene Aliases
-
Chromosome Location
Chr.7: 34109541 - 34133525 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207212.2 NP_997095.1 2 - 3 883 79
GenBank mRNA BC042762.1 - 2 - 3 315 79
BC054125.1 - 2 - 3 883 79

View Full Product Details