The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026549.3 | NP_080825.1 | 5 - 6 | 817 | 107 |
GenBank mRNA | AK003339.1 | - | 5 - 6 | 826 | 107 |
AK077948.1 | - | 5 - 6 | 818 | 107 | |
AK151495.1 | - | 5 - 6 | 821 | 107 | |
AK154811.1 | - | 5 - 6 | 833 | 107 | |
BC025090.1 | - | 2 - 3 | 281 | 107 | |
BC038524.1 | - | 2 - 3 | 287 | 107 | |
BC115797.1 | - | 5 - 6 | 816 | 107 |