Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Fam45a
Entrez Gene ID
67894
Gene Name
family with sequence similarity 45, member A
Gene Aliases
1810055E12Rik
Chromosome Location
Chr.19: 60811441 - 60836235 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK171767.1 - 3 - 3 90

View Full Product Details