The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_207261.3 | NP_997144.1 | 3 - 3 | 1502 | 69 |
GenBank mRNA | AK139600.1 | - | 3 - 3 | 1936 | 69 |
AK162136.1 | - | 3 - 3 | 1505 | 69 |