The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001113559.2 | NP_001107031.2 | 14 - 14 | 7141 | 118 |
NM_001243163.1 | NP_001230092.1 | 14 - 14 | 7033 | 118 | |
NM_011444.3 | NP_035574.2 | 15 - 15 | 7285 | 118 | |
GenBank mRNA | AK031243.1 | - | 1 - 1 | 2191 | 119 |