Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Casp7
Entrez Gene ID
12369
Gene Name
caspase 7
Gene Aliases
AI314680, CMH-1, ICE-IAP3, Mch3, caspase-7, mCASP-7
Chromosome Location
Chr.19: 56396091 - 56442347 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_007611.2 NP_031637.1 1 - 2 191 121

View Full Product Details