The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177732.4 | NP_808400.1 | 1 - 2 | 218 | 69 |
GenBank mRNA | AK049180.1 | - | 1 - 2 | 219 | 69 |
AK049462.1 | - | 1 - 2 | 138 | 69 | |
AK052968.1 | - | 1 - 2 | 138 | 69 | |
AK140305.1 | - | 1 - 2 | 510 | 69 | |
AK164517.1 | - | 1 - 2 | 138 | 69 | |
AK169775.1 | - | 1 - 2 | 120 | 69 | |
AK220330.1 | - | 1 - 2 | 128 | 69 | |
BC115984.1 | - | 2 - 3 | 119 | 69 |