Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dmrtb1
Entrez Gene ID
56296
Gene Name
DMRT-like family B with proline-rich C-terminal, 1
Gene Aliases
Dmrt6, Prp13
Chromosome Location
Chr.4: 107676288 - 107684243 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019872.1 NP_063925.1 2 - 3 796 63
GenBank mRNA AF020191.1 - 1 - 2 228 63
AK132787.1 - 2 - 3 326 63
AK161436.1 - 2 - 3 306 63

View Full Product Details