The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001205156.1 | NP_001192085.1 | 8 - 9 | 1022 | 68 |
NM_145550.3 | NP_663525.1 | 9 - 10 | 1103 | 68 | |
GenBank mRNA | AI303400.1 | - | 1 - 2 | 68 | |
AK081205.1 | - | 9 - 10 | 1090 | 68 | |
AK157536.1 | - | 9 - 10 | 1090 | 68 | |
BC009080.1 | - | 9 - 10 | 1035 | 68 |