The assay primers and probes lie within a single exon. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011033.2 | NP_035163.1 | 1 - 1 | 1956 | 66 |
GenBank mRNA | AK077154.1 | - | 1 - 1 | 1956 | 66 |
BC051134.1 | - | 1 - 1 | 1936 | 66 | |
CN837699.1 | - | 1 - 1 | 116 | 66 | |
X75959.1 | - | 1 - 1 | 1932 | 66 |