Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc2a7
Entrez Gene ID
435818
Gene Name
solute carrier family 2 (facilitated glucose transporter), member 7
Gene Aliases
OTTMUSG00000010396
Chromosome Location
Chr.4: 150148837 - 150168895 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085529.1 NP_001078998.1 1 - 2 49 70

View Full Product Details