The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001002272.3 | NP_001002272.1 | 11 - 12 | 2433 | 92 |
NM_001290770.1 | NP_001277699.1 | 11 - 12 | 2382 | 92 | |
NM_001290771.1 | NP_001277700.1 | 11 - 12 | 2433 | 92 | |
NM_019548.4 | NP_062421.2 | 11 - 12 | 2433 | 92 | |
GenBank mRNA | AB017108.1 | - | 9 - 10 | 774 | 92 |
AB032477.1 | - | 11 - 12 | 2301 | 92 | |
AB036783.1 | - | 12 - 13 | 2375 | 92 | |
AF241244.1 | - | 11 - 12 | 2297 | 92 | |
AF241245.1 | - | 11 - 12 | 2297 | 92 | |
AF288605.1 | - | 12 - 13 | 2375 | 92 | |
AF319977.1 | - | 10 - 11 | 2316 | 92 | |
AK122449.1 | - | 10 - 11 | 1323 | 92 | |
BC075630.1 | - | 11 - 12 | 2382 | 92 |