Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Tro
Entrez Gene ID
56191
Gene Name
trophinin
Gene Aliases
AA409408, Maged3, Maged3l, Tnn, Trol, magphinin
Chromosome Location
Chr.X: 150644710 - 150657583 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001002272.3 NP_001002272.1 2 - 3 168 80
NM_001290770.1 NP_001277699.1 2 - 3 117 80
NM_001290771.1 NP_001277700.1 2 - 3 168 80
NM_001290772.1 NP_001277701.1 2 - 3 168 80
NM_019548.4 NP_062421.2 2 - 3 168 80
GenBank mRNA AB036783.1 - 2 - 3 131 80
AF241244.1 - 1 - 2 53 80
AF241245.1 - 1 - 2 53 80
AF288605.1 - 2 - 3 131 80
AF288606.1 - 2 - 3 131 80
AF319977.1 - 1 - 2 51 80
AK047777.1 - 2 - 3 192 80
BC075630.1 - 2 - 3 117 80

View Full Product Details