Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Xpnpep1
Entrez Gene ID
170750
Gene Name
X-prolyl aminopeptidase (aminopeptidase P) 1, soluble
Gene Aliases
D230045I08Rik, sAMP
Chromosome Location
Chr.19: 52932227 - 53038654 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133216.3 NP_573479.3 6 - 7 615 75
GenBank mRNA AF363970.1 - 6 - 7 555 75
AK146033.1 - 6 - 7 595 75
AK149676.1 - 6 - 7 554 75
AK158597.1 - 4 - 5 573 75
AK166715.1 - 6 - 7 515 75
AK169024.1 - 5 - 6 441 75
BC065174.1 - 6 - 7 503 75

View Full Product Details