Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pex11b
Entrez Gene ID
18632
Gene Name
peroxisomal biogenesis factor 11 beta
Gene Aliases
PEX11beta, Pex11pbeta
Chromosome Location
Chr.3: 96635357 - 96645381 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_027845.1 1 - 2 151 62
GenBank mRNA AK082778.1 - 1 - 2 42 62
BQ443231.1 - 1 - 2 131 62
BY285331.1 - 1 - 2 150 62

View Full Product Details