Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Pex11b
Entrez Gene ID
18632
Gene Name
peroxisomal biogenesis factor 11 beta
Gene Aliases
PEX11beta, Pex11pbeta
Chromosome Location
Chr.3: 96635357 - 96645381 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001162387.1 NP_001155859.1 2 - 3 432 71
NM_001162388.1 NP_001155860.1 2 - 3 196 71
NM_011069.3 NP_035199.3 2 - 3 432 71
NR_027844.1 2 - 3 428 71
NR_027845.1 2 - 3 262 71
GenBank mRNA AF093671.1 - 2 - 3 177 71
AK005236.1 - 2 - 3 184 71
AK011709.1 - 2 - 3 230 71
AK082279.1 - 2 - 3 191 71
AK082778.1 - 2 - 3 153 71
BC013812.1 - 2 - 3 181 71
BC054414.1 - 2 - 3 180 71
BQ443231.1 - 2 - 3 242 71
BY285331.1 - 2 - 3 72
CD806660.1 - 2 - 3 422 71

View Full Product Details