The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033172.3 | NP_001028344.2 | 3 - 4 | 1623 | 70 |
NM_001164367.1 | NP_001157839.1 | 3 - 4 | 1051 | 70 | |
GenBank mRNA | AK015534.1 | - | 3 - 4 | 1051 | 70 |
AK154916.1 | - | 3 - 4 | 1623 | 70 | |
BC089010.1 | - | 2 - 3 | 882 | 70 | |
BC139380.1 | - | 3 - 4 | 1386 | 70 | |
BC139419.1 | - | 3 - 4 | 1386 | 70 |