Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fam45a
Entrez Gene ID
67894
Gene Name
family with sequence similarity 45, member A
Gene Aliases
1810055E12Rik
Chromosome Location
Chr.19: 60811441 - 60836235 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001167829.1 NP_001161301.1 2 - 3 302 114
NM_026437.3 NP_080713.2 2 - 3 302 114
GenBank mRNA AK007867.1 - 2 - 3 262 114
AK145769.1 - 2 - 3 268 114
BC013514.1 - 2 - 3 261 114

View Full Product Details