The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019946.4 | NP_064330.2 | 1 - 2 | 70 | 63 |
GenBank mRNA | AK002800.1 | - | 1 - 2 | 64 | |
AK005122.1 | - | 1 - 2 | 71 | 63 |