Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bcl9
Entrez Gene ID
77578
Gene Name
B cell CLL/lymphoma 9
Gene Aliases
2610202E01Rik, 8030475K17Rik, A330041G23Rik, Gm130
Chromosome Location
Chr.3: 97203658 - 97298083 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029933.4 NP_084209.3 2 - 3 280 72
GenBank mRNA AK133757.1 - 2 - 3 272 72
AK147659.1 - 2 - 3 280 72
BC022939.1 - 2 - 3 218 72

View Full Product Details