Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
March3
Entrez Gene ID
320253
Gene Name
membrane-associated ring finger (C3HC4) 3
Gene Aliases
6330411I15Rik, A530081L18Rik, AI843639, MARCH-III
Chromosome Location
Chr.18: 56761716 - 56925652 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_177115.2 NP_796089.1 1 - 2 240 87
GenBank mRNA AK036622.1 - 1 - 2 208 87
AK041088.1 - 1 - 2 240 87

View Full Product Details