Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Creb1
- Entrez Gene ID
-
12912
- Gene Name
- cAMP responsive element binding protein 1
- Gene Aliases
-
2310001E10Rik, 3526402H21Rik, AV083133, Creb, Creb-1
- Chromosome Location
-
Chr.1: 64532804 - 64604548 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
U46027.1 |
- |
4 - 5 |
485 |
98 |
View Full Product Details