Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pnkd
Entrez Gene ID
56695
Gene Name
paroxysmal nonkinesiogenic dyskinesia
Gene Aliases
2210013N15Rik, 2810403H05Rik, AI854243, Brp17, MNCb-5687, MR-1, Tahccp2
Chromosome Location
Chr.1: 74284597 - 74353692 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001039509.1 NP_001034598.1 2 - 3 271 61
GenBank mRNA AK173125.1 - 2 - 3 245 61

View Full Product Details