Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Asnsd1
Entrez Gene ID
70396
Gene Name
asparagine synthetase domain containing 1
Gene Aliases
2210409M21Rik
Chromosome Location
Chr.1: 53344617 - 53352752 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001290984.1 NP_001277913.1 4 - 5 463 93
GenBank mRNA AK168534.1 - 4 - 5 464 93

View Full Product Details