Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankzf1
Entrez Gene ID
52231
Gene Name
ankyrin repeat and zinc finger domain containing 1
Gene Aliases
1300008P06Rik, 2810025E10Rik, D1Ertd161e
Chromosome Location
Chr.1: 75192160 - 75199387 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_051999.1 1 - 2 521 60
GenBank mRNA AK084573.1 - 1 - 2 483 60

View Full Product Details