Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Srsf2
- Entrez Gene ID
-
20382
- Gene Name
- serine/arginine-rich splicing factor 2
- Gene Aliases
-
D11Wsu175e, MRF-1, Pr264, SC35, Sfrs10, Sfrs2
- Chromosome Location
-
Chr.11: 116849897 - 116853104 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BC005493.1 |
- |
2 - 3 |
819 |
109 |
View Full Product Details