Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc14a2
Entrez Gene ID
27411
Gene Name
solute carrier family 14 (urea transporter), member 2
Gene Aliases
UT-A1, UT-A3
Chromosome Location
Chr.18: 78146144 - 78596950 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001110273.1 NP_001103743.1 4 - 5 1058 79
NM_207651.3 NP_997534.2 15 - 16 2198 79
GenBank mRNA AF366052.2 - 15 - 16 2190 79
AF367359.1 - 4 - 5 1019 79
AK032837.1 - 6 - 7 802 79
BC150680.1 - 15 - 16 2106 79

View Full Product Details