Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Pqlc1
Entrez Gene ID
66943
Gene Name
PQ loop repeat containing 1
Gene Aliases
2310009N05Rik, 4933425L21Rik, 5730564E11Rik, C78974
Chromosome Location
Chr.18: 80255241 - 80292724 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_025861.3 NP_080137.2 4 - 5 625 87
GenBank mRNA AK018759.1 - 3 - 4 1003 87
BC043686.1 - - 803 87

View Full Product Details