Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Htr4
- Entrez Gene ID
-
15562
- Gene Name
- 5 hydroxytryptamine (serotonin) receptor 4
- Gene Aliases
-
5-HT-4, 5-HT4, 5-HT<4L>, 5HTR4
- Chromosome Location
-
Chr.18: 62322160 - 62498026 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
AJ011369.1 |
- |
5 - 6 |
1076 |
77 |
View Full Product Details