Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ccbe1
Entrez Gene ID
320924
Gene Name
collagen and calcium binding EGF domains 1
Gene Aliases
4933426F18Rik, 9430093N24Rik, mKIAA1983
Chromosome Location
Chr.18: 66056855 - 66291838 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178793.4 NP_848908.1 3 - 4 345 70
GenBank mRNA AK028377.1 - 3 - 4 343 70
AK035153.1 - 3 - 4 340 70
AK039742.1 - 3 - 4 275 70
BC103803.1 - 3 - 4 295 70
BC152322.1 - 3 - 4 314 70

View Full Product Details