Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Nars
Entrez Gene ID
70223
Gene Name
asparaginyl-tRNA synthetase
Gene Aliases
3010001M15Rik, AA960128, ASNRS, C78150
Chromosome Location
Chr.18: 64499647 - 64516557 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001142950.1 NP_001136422.1 3 - 4 287 109
NM_027350.3 NP_081626.2 3 - 4 284 109
GenBank mRNA AK083730.1 - 3 - 4 244 109
AK157244.1 - 3 - 4 246 109
AK172657.1 - 3 - 4 246 109
AK191424.1 - 2 - 3 105 109
BC005473.1 - 3 - 4 190 109
BC052849.1 - 3 - 4 245 109

View Full Product Details