The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027350.3 | NP_081626.2 | 1 - 2 | 140 | 78 |
GenBank mRNA | AK077699.1 | - | 1 - 2 | 121 | 78 |