Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Smad2
Entrez Gene ID
17126
Gene Name
SMAD family member 2
Gene Aliases
7120426M23Rik, Madh2, Madr2, Smad-2, mMad2
Chromosome Location
Chr.18: 76241251 - 76311748 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AY334552.1 - 1 - 2 236 81
BU058005.1 - 2 - 3 598 81

View Full Product Details