Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mc2r
Entrez Gene ID
17200
Gene Name
melanocortin 2 receptor
Gene Aliases
ACTH-R, ACTHR, MC2-R
Chromosome Location
Chr.18: 68406899 - 68429320 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001271716.1 NP_001258645.1 3 - 4 271 100
NM_001271717.1 NP_001258646.1 2 - 3 183 100
NM_001301372.1 NP_001288301.1 4 - 5 357 100
NM_008560.3 NP_032586.1 2 - 3 214 100
GenBank mRNA AK033100.1 - 2 - 3 153 100
BC139210.1 - 1 - 2 117 100
BC139211.1 - 1 - 2 116 100
D85688.1 - 3 - 4 271 100

View Full Product Details