The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001170960.1 | NP_001164431.1 | 3 - 4 | 505 | 86 |
NM_026854.3 | NP_081130.1 | 5 - 6 | 793 | 86 | |
GenBank mRNA | AK004450.1 | - | 5 - 6 | 768 | 86 |
AK144689.1 | - | 5 - 6 | 768 | 86 | |
BC139347.1 | - | 5 - 6 | 759 | 86 | |
BC139348.1 | - | 5 - 6 | 759 | 86 | |
BC145608.1 | - | 4 - 5 | 566 | 86 | |
BC145609.1 | - | 3 - 4 | 471 | 86 |