Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mcc
Entrez Gene ID
328949
Gene Name
mutated in colorectal cancers
Gene Aliases
D18Ertd451e, E330037C19
Chromosome Location
Chr.18: 44425060 - 44812182 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001085373.1 NP_001078842.1 6 - 7 999 72
NM_001085374.1 NP_001078843.1 4 - 5 967 72
GenBank mRNA AK054528.1 - 4 - 5 969 72
AK158266.1 - 6 - 7 1000 72

View Full Product Details