Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dctn4
Entrez Gene ID
67665
Gene Name
dynactin 4
Gene Aliases
1110001K06Rik, 4930547K17Rik, C130039E17Rik, p62
Chromosome Location
Chr.18: 60526159 - 60558762 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK034292.1 - 5 - 6 589 79
AK141855.1 - 5 - 6 553 79

View Full Product Details