Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Astn2
Entrez Gene ID
56079
Gene Name
astrotactin 2
Gene Aliases
1d8, Astnl, bM452J22.1
Chromosome Location
Chr.4: 65380803 - 66404530 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019514.3 NP_062387.2 17 - 18 3210 87
NM_207109.2 NP_996992.1 18 - 19 3366 87
GenBank mRNA AF116573.1 - 15 - 16 2378 87
AK041160.1 - 19 - 20 3270 87

View Full Product Details