Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rfx5
Entrez Gene ID
53970
Gene Name
regulatory factor X, 5 (influences HLA class II expression)
Gene Aliases
-
Chromosome Location
Chr.3: 94951957 - 94961561 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA BC075717.1 - 2 - 3 115 87

View Full Product Details