The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_175471.2 | NP_780680.1 | 5 - 6 | 929 | 64 |
GenBank mRNA | AK053031.1 | - | 8 - 9 | 1334 | 64 |
BC004750.1 | - | 4 - 5 | 663 | 64 | |
BC043687.1 | - | 5 - 6 | 929 | 64 |