Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- Trp73
- Entrez Gene ID
-
22062
- Gene Name
- transformation related protein 73
- Gene Aliases
-
TAp73, Tp73, deltaNp73, p73
- Chromosome Location
-
Chr.4: 154056249 - 154140208 on Build GRCm38
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
GenBank mRNA |
BB663805.1 |
- |
1 - 2 |
129 |
89 |
View Full Product Details