Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Mfsd11
Entrez Gene ID
69900
Gene Name
major facilitator superfamily domain containing 11
Gene Aliases
2600014M03Rik
Chromosome Location
Chr.11: 116852283 - 116875660 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_178620.3 NP_848735.1 3 - 4 320 87
GenBank mRNA AK031384.1 - 3 - 4 515 87
BC088988.1 - 3 - 4 320 87
BC137970.1 - 3 - 4 307 87
BC137971.1 - 3 - 4 308 87

View Full Product Details