Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Fbxo6
Entrez Gene ID
50762
Gene Name
F-box protein 6
Gene Aliases
AA408845, FBG2, Fbs2, Fbx6b, Fbxo6b
Chromosome Location
Chr.4: 148145716 - 148152135 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001163707.1 NP_001157179.1 1 - 2 124 103
GenBank mRNA BY164736.1 - 1 - 2 129 103

View Full Product Details