Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tprgl
Entrez Gene ID
67808
Gene Name
transformation related protein 63 regulated like
Gene Aliases
1200015A19Rik, Svap30, Tprg1l, mover
Chromosome Location
Chr.4: 154157485 - 154160712 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK178710.1 - 1 - 2 56 77
AK180396.1 - 1 - 2 56 78
AK214091.1 - 1 - 2 56 77

View Full Product Details