The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001039157.2 | NP_001034246.1 | 5 - 6 | 643 | 70 |
NM_001039158.2 | NP_001034247.1 | 4 - 5 | 617 | 70 | |
NM_001039159.2 | NP_001034248.1 | 4 - 5 | 480 | 70 | |
NM_001284312.1 | NP_001271241.1 | 5 - 6 | 544 | 70 | |
NM_001284314.1 | NP_001271243.1 | 4 - 5 | 480 | 70 | |
NM_021402.6 | NP_067377.4 | 5 - 6 | 780 | 70 | |
GenBank mRNA | AF296656.1 | - | 3 - 4 | 300 | 70 |
AK004626.1 | - | 4 - 5 | 464 | 70 | |
AK076265.1 | - | 5 - 6 | 745 | 70 | |
AW826087.1 | - | 4 - 5 | 301 | 70 | |
BC065779.1 | - | 5 - 6 | 544 | 70 | |
U93242.1 | - | 3 - 4 | 300 | 70 |