Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Espn
Entrez Gene ID
56226
Gene Name
espin
Gene Aliases
je
Chromosome Location
Chr.4: 152120331 - 152152420 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_207687.2 NP_997570.1 9 - 10 1752 143
NM_207688.2 NP_997571.1 4 - 5 781 143
NM_207691.2 NP_997574.2 4 - 5 781 143
GenBank mRNA AF239886.1 - 9 - 10 1752 143
AF540942.1 - 4 - 5 781 143
AF540943.1 - 4 - 5 781 143
AK145160.1 - 2 - 3 316 143
AK146701.1 - 2 - 3 281 143
BY739473.1 - 2 - 3 281 143

View Full Product Details