Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Slc38a6
Entrez Gene ID
625098
Gene Name
solute carrier family 38, member 6
Gene Aliases
AW322671, EG625098
Chromosome Location
Chr.12: 73286794 - 73361998 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001037717.3 NP_001032806.2 15 - 16 1368 109
GenBank mRNA BC043932.1 - 8 - 9 695 109
BC157917.1 - 15 - 16 1317 109
BC157928.1 - 15 - 16 1317 109

View Full Product Details