The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001033490.1 | NP_001028662.1 | 2 - 3 | 195 | 70 |
GenBank mRNA | AK160236.1 | - | 1 - 2 | 359 | 70 |